Learn More
MDC Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P176100UG

Description
AA Sequence of recombinant protein: GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MDC (CCL22). Anti-Murine MDC (CCL22)-specific antibody was purified by affinity chromatography employing an immobilized Murine MDC (CCL22) matrix. Sandwich ELISA: To detect Murine MDC (CCL22) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine MDC (CCL22) (500-P176Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MDC (CCL22). Western Blot: To detect Murine MDC (CCL22) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine MDC (CCL22) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
MDC Polyclonal antibody specifically detects MDC in Mouse samples. It is validated for ELISA, Western BlotSpecifications
MDC | |
Polyclonal | |
PBS with no preservative | |
O88430 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20299 | |
-20°C |
ELISA, Western Blot | |
Unconjugated | |
CCL22 | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
E.coli-derived Recombinant Murine MDC (CCL22) | |
100 μg | |
Primary | |
Mouse | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.