Learn More
MDGA2, Mouse anti-Human,Rat, Alexa Fluor 350, Clone: 803732, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB5184U100UG
Catalog No. FAB5184U100
Add to cart
This item is not returnable. View return policy
Description
Description
MDGA2 Monoclonal specifically detects MDGA2 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
MDGA2
803732
Flow Cytometry 0.25-1 ug/10^6 cells
Q7Z553
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1
Mouse myeloma cell line NS0-derived recombinant human MDGA2, Gln21-Asp934, Accession # Q7Z553
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
161357
Human, Rat
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human MDGA2 in direct ELISAs. Detects rat MDGA2 in Flow Cytometry.