Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH1 Mouse anti-Human, Clone: CPTC-MDH1-1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen MDH1
Clone CPTC-MDH1-1
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122051
View Documents
Novus Biologicals
NBP30728910UG
100 μg
Each for $352.89
Add to cart
 
NB122052
View Documents
Novus Biologicals
NBP30728920UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

MDH1 Monoclonal antibody specifically detects MDH1 in Human samples. It is validated for Western Blot
Specifications

Specifications

MDH1
0.2 mg/ml
Western Blot
Purified
RUO
Human
cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase
Recombinant human full-length MDH1 protein
Monoclonal
Protein A or G purified
CPTC-MDH1-1
Western Blot 1-2 ug/ml
Unconjugated
Mouse
Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
4190
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit