Learn More
MDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159041AP
Catalog No. 50-173-0564
Add to cart
Description
Description
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Specifications
Specifications
MDH1
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88989, P14152, P40925
Cytosolic malate dehydrogenase, MDH s, MDH1, MDHA, MOR2
Rabbit
IgG
150 μL
-20°C
Primary
17449, 24551, 4190
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
MDH1
Liquid
Mdh1
MDH1 Fusion Protein Ag8744
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat