Learn More
Aviva Systems Biology Corp MDH1B Antibody - middle region (ARP51858_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDH1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MDH1B. Alternate names: FLJ25341, RP11-95H11ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62843