Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDL 28170, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
88191-84-8
C22H26N2O4
382.46
NGBKFLTYGSREKK-WOJBJXKFSA-N
calpain inhibitor iii,benzyl n-1s-2-methyl-1-1-oxo-3-phenylpropan-2-yl carbamoyl propyl carbamate,1-1-formyl-2-phenyl-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,mdl hplc,n-n-phenylmethoxy carbonyl-l-valyl-phenylalaninal,s-2-s-2-benzyloxycarbonylamino-3-methyl-butyrylamino-3-phenyl-propionic acid,s-1-1-benzyl-2-oxo-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,1-1-benzyl-2-oxo-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,benzyl 2s-3-methyl-1-oxo-1-1-oxo-3-phenylpropan-2-ylamino butan-2-ylcarbamate,benzyl s-3-methyl-1-oxo-1-1-oxo-3-phenylpropan-2-ylamino butan-2-ylcarbamate
51435906
benzyl N-[(2R)-3-methyl-1-oxo-1-[[(2R)-1-oxo-3-phenylpropan-2-yl]amino]butan-2-yl]carbamate
CC(C)C(C(=O)NC(CC1=CC=CC=C1)C=O)NC(=O)OCC2=CC=CC=C2
88191-84-8
C22H26N2O4
382.46
NGBKFLTYGSREKK-WOJBJXKFSA-N
calpain inhibitor iii,benzyl n-1s-2-methyl-1-1-oxo-3-phenylpropan-2-yl carbamoyl propyl carbamate,1-1-formyl-2-phenyl-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,mdl hplc,n-n-phenylmethoxy carbonyl-l-valyl-phenylalaninal,s-2-s-2-benzyloxycarbonylamino-3-methyl-butyrylamino-3-phenyl-propionic acid,s-1-1-benzyl-2-oxo-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,1-1-benzyl-2-oxo-ethylcarbamoyl-2-methyl-propyl-carbamic acid benzyl ester,benzyl 2s-3-methyl-1-oxo-1-1-oxo-3-phenylpropan-2-ylamino butan-2-ylcarbamate,benzyl s-3-methyl-1-oxo-1-1-oxo-3-phenylpropan-2-ylamino butan-2-ylcarbamate
51435906
benzyl N-[(2R)-3-methyl-1-oxo-1-[[(2R)-1-oxo-3-phenylpropan-2-yl]amino]butan-2-yl]carbamate
CC(C)C(C(=O)NC(CC1=CC=CC=C1)C=O)NC(=O)OCC2=CC=CC=C2

Potent, selective calpain and cathepsin B inhibitor

Potent, selective inhibitor of calpain and cathepsin B (K i values are 10 and 25 nM respectively) that does not inhibit trypsin-like serine proteases. Rapidly penetrates the blood-brain barrier following systemic administration and displays neuroprotective effects in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 382.45
Percent Purity >95%
Chemical Name or Material MDL 28170

Recommended Storage : Store at -20°C