Tocris™ MDL 72527

Polyamine oxidase (PAO) inhibitor

Manufacturer: Tocris™ 3709/50

 View more versions of this product

Catalog No. 37-095-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors MDL 72527
    Molecular Weight 265.23
    Quantity 50mg
    Product Type MDL 72527

    Polyamine oxidase (POA) inhibitor. Does not inhibit monoamine oxidase or D-Amino acid oxidase. Displays anticancer and neuroprotective activity in vivo.

    • Chemical Name: N1,N4-Di-2,3-butadienyl-1,4-butanediamine dihydrochloride
    • Formula: C12H20N2.2HCl
    • CAS Number: 93565-01-6
    • Solubility: Soluble to 100mM in water and to 75mM in DMSO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit