Learn More
Tocris Bioscience™ MDL 72832 hydrochloride
Potent 5-HT1A ligand
Supplier: Tocris Bioscience™ 0412/50
This item is not returnable.
View return policy
Description
MDL 72832 hydrochloride is a potent ligand at 5-HT1A receptors, with mixed agonist and antagonist properties. An agonist at post-synaptic 5-HT1A receptors.Specifications
8-[4-(1,4-Benzodioxan-2-ylmethylamino)butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride | |
>99% | |
Soluble to 5 mM in water | |
Observed MW: 422.95 |
113777-40-5 | |
50 mg | |
C22H30N2O4.HCl |
Safety and Handling
Recommended Storage : Store at Room Temperature