Learn More
MDR1/ABCB1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MRK16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252701AF350
Catalog No. NP22701A30
Add to cart
Description
Description
MDR1/ABCB1 Monoclonal antibody specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry,Immunofluorescence.
Specifications
Specifications
MDR1/ABCB1
MRK16
Flow Cytometry, Immunofluorescence
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Recognizes a discontinuous extracellular epitope on P-glycoprotein.
Flow Cytometry, Immunofluorescence
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ABCB1
Drug resistant K-562 cells.
0.1 mL
Primary
5243
Human