Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1/ABCB1 Mouse anti-Human, Alexa Fluor™ 594, Clone: MRK16, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252701AF594

Catalog No. NP22701A94


Add to cart

Description

Description

MDR1/ABCB1 Monoclonal antibody specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

MDR1/ABCB1
MRK16
Flow Cytometry, Immunofluorescence
Conjugated, Purified
ABCB1
Drug resistant K-562 cells.
Protein A or G purified
Cancer, Membrane Trafficking and Chaperones
Primary
5243.0
Human
Flow Cytometry, Immunofluorescence
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Recognizes a discontinuous extracellular epitope on P-glycoprotein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit