Learn More
MDR1/ABCB1 Mouse anti-Human, DyLight 680, Clone: 6G11F8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237290FR
Catalog No. NBP237290FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MDR1/ABCB1
6G11F8
Western Blot, ELISA
Purified
ABCB1
Purified recombinant fragment of human ABCB1 (AA: 632-693) expressed in E. Coli.
Protein A or G purified
Cancer, Membrane Trafficking and Chaperones
Primary
5243
Western Blot, ELISA
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human