Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1/ABCB1 Mouse anti-Human, FITC, Clone: 6G11F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237290F

Catalog No. NBP237290F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MDR1/ABCB1
6G11F8
ELISA
Purified
ABCB1
Purified recombinant fragment of human ABCB1 (AA: 632-693) expressed in E. Coli.
Protein A or G purified
Cancer, Membrane Trafficking and Chaperones
Primary
5243
ELISA
FITC
PBS with 0.05% Sodium Azide
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit