ME 0328, Tocris™

Manufacturer: R&D Systems 5054/10

Quantity 10mg

 View more versions of this product

Catalog No. 505410R

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1445251-22-8
    Physical Form Solid
    Assay Percent Range >98%
    Molecular Formula C19H19N3O2
    Formula Weight 321.37
    Chemical Name or Material ME 0328

    Specifications

    Physical Form Solid
    Formula Weight 321.37
    Color White
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C19H19N3O2
    CAS 1445251-22-8
    Recommended Storage Store at +4°C
    Chemical Name or Material ME 0328

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit