Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ME1 Antibody (736), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243709
Description
Ensure accurate, reproducible results in Western Blot
ME1 Monoclonal specifically detects ME1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ME1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ME1 | |
| Recombinant protein encompassing a sequence within the center region of human ME1. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| 4199 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 736 | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| EC 1.1.1, EC 1.1.1.40, HUMNDME, malate dehydrogenase, Malic enzyme 1, malic enzyme 1, NADP(+)-dependent, cytosolic, malic enzyme 1, soluble, Malic enzyme, cytoplasmic, MES, NADP-dependent malic enzyme, NADP-ME, pyruvic-malic carboxylase | |
| Mouse | |
| 64 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction