Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ME1 Mouse anti-Human, Clone: GT736, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527762

Catalog No. PIMA527762


Add to cart

Description

Description

Positive Control: 293T, A431, HeLa Predicted Reactivity: Mouse (91%), Rat (91%), Chicken (80%), Sheep (92%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.
Specifications

Specifications

ME1
Monoclonal
1 mg/ml
PBS with no preservative
P48163
MES, malic enzyme 1, NADP(+)-dependent, cytosolic, HUMNDME
Mouse
IgG2b
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4199
Immunocytochemistry, Immunofluorescence, Western Blot
GT736
Unconjugated
ME1
Liquid
ME1
Recombit protein encompassing a sequence within the center region of human ME1. The exact sequence is proprietary.
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit