Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ME1 Mouse anti-Human, Mouse, DyLight 488, Clone: 979, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243710G

Catalog No. NBP243710G


Add to cart

Description

Description

ME1 Monoclonal antibody specifically detects ME1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ME1
979
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
ME1
Recombinant protein encompassing a sequence within the center region of human ME1. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 1.1.1, EC 1.1.1.40, HUMNDME, malate dehydrogenase, Malic enzyme 1, malic enzyme 1, NADP(+)-dependent, cytosolic, malic enzyme 1, soluble, Malic enzyme, cytoplasmic, MES, NADP-dependent malic enzyme, NADP-ME, pyruvic-malic carboxylase
Mouse
IgG1
0.1 ml
Primary
4199
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit