Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mecamylamine hydrochloride, Tocris Bioscience™

Catalog Number p-4784135
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
110691-49-1
C11H22ClN
203.754
PKVZBNCYEICAQP-CIISUUNXSA-N
mecamylamine hcl, 2-methylamino isocamphane hydrochloride, n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, a: 600/1 223m072 melphalan hydrochloride 3223-07-2, 1s,4r-n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, mecamylamine hydrochloride, united states pharmacopeia usp reference standard
25162921
(1R,4S)-N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
CC1(C2CCC(C2)C1(C)NC)C.Cl
This item is not returnable. View return policy
110691-49-1
C11H22ClN
203.754
PKVZBNCYEICAQP-CIISUUNXSA-N
mecamylamine hcl, 2-methylamino isocamphane hydrochloride, n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, a: 600/1 223m072 melphalan hydrochloride 3223-07-2, 1s,4r-n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, mecamylamine hydrochloride, united states pharmacopeia usp reference standard
25162921
(1R,4S)-N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
CC1(C2CCC(C2)C1(C)NC)C.Cl

Non-competitive nAChR antagonist

Non-competitive nicotinic acetylcholine receptor antagonist. Displays antidepressant-like effects in mice.
Quantity 10 mg
Formula Weight 203.75
Percent Purity >98%
Chemical Name or Material (1 R,2 S,4 S)-rel-N,2,3,3-Tetramethylbicyclo[2.2.1 ]heptan-2-amine hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit