Mechanix Wear™ FastFit™ Mechanics Gloves

Multipurpose glove suited for a variety of industrial and maintenance activities requiring solid handling and dexterity

Manufacturer: Mechanix Wear MFF03010

 View more versions of this product

Catalog No. 19-030-019C

  Check Availability
Add to Cart

    Specifications

    Finger Style Full
    Exterior Finish Smooth
    Glove Cuff Style Shirred
    For Use With (Application) variety of industrial and maintenance activities requiring solid handling and dexterity
    Material Synthetic Leather, Spandex™
    Liner Material Spandex™
    Size Large
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit