Learn More
Mechanix Wear™ FastFit™ Red Mechanics Gloves
These high-dexterity, synthetic leather gloves are touchscreen compatible to help users stay connected while on the job. FastFit™ gloves offer ideal fit with TrekDry™ evaporative cooling technology to keep hands cool and comfortable.
Manufacturer:  Mechanix Wear™ MFF02010
 View more versions of this product
Catalog No. 19-030-018C
Add to cart
This item is not returnable. View return policy
Description
Description
  • TrekDry™ material is form-fitting to keep working hands cool and comfortable
  • High-dexterity synthetic leather infused with touchscreen technology
  • Stretch-elastic cuffs create a secure fit
  • Reinforced thumb and index finger provide additional durability
  • Anatomically designed two-piece palm design prevents material bunching
  • Pinched fingertip construction improves fingertip strength and durability
  • Machine washable
Specifications
Specifications
TrekDry™
Uncoated
Specific
Mechanics Gloves
Maintenance Repairs, Equipment Operation, and DIY Home Improvement
FastFit™
Elastic
Red
Synthetic Leather
Large
Open