Mechanix Wear™ M-Pact 2 Gloves
Extensive padding provides comfort and stands up to high-impact abuse
Manufacturer: Mechanix WearSupplier Diversity Partner MP201009
 View all options for this product
Catalog No. 19-030-021B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Material Synthetic Leather
    Size Medium
    Double layer synthetic leather palm is cushioned by an anatomically die-cut foam-padded layer; wrist, top panel, and fingers are all covered by a molded thermal plastic rubber (TPR) shock-absorbing pad; and fingertips and thumb panel are reinforced for reduced wear.
    ·     High-density accordion-style surface blunts pointed impacts to the knuckle panel
    ·     Side panels feature synthetic leather finger panels with breathable Lycra™ inserts
    ·     Enlarged entry for easy access
    ·     Adjustable Spandex™ stretch wrist panel insert helps to seal out dirt
    ·     Wide-band elastic cuff, hook and loop closure, and TPR pull-tab for a secure fit
    Machine washable. Black.
  • Description & Specifications
Related Products
Product Content Feedback