Learn More
Mechanix Wear™ M-Pact™ Covert Tactical Impact Gloves
Durable, 0.8 mm synthetic leather construction infused with touchscreen technology for convenient use with smart devices. D3O™ palm padding dissipates high-impact energy and reduces hand fatigue during military or law enforcement applications.
$83.10
Specifications
Color | Black |
---|---|
Glove Cuff Style | Hook and Loop |
Hand | Specific |
Material | Synthetic Leather, Thermoplastic Rubber |
Palm Type | D3O™ Padding |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-030-016A
|
Mechanix Wear™
MPT55008 |
Small |
Pair for $83.10
|
|
19-030-016B
|
Mechanix Wear™
MPT55009 |
Medium |
Pair for $83.10
|
|
19-030-016C
|
Mechanix Wear™
MPT55010 |
Large |
Pair for $83.10
|
|
19-030-016D
|
Mechanix Wear™
MPT55011 |
X-Large |
Pair for $83.10
|
|
19-030-016E
|
Mechanix Wear™
MPT55012 |
2X-Large |
Pair for $83.10
|
|
Description
- Thermoplastic Rubber (TPR) provides a high level of impact protection
- Breathable TrekDry™ keeps hands cool and comfortable
- Armortex™ palm reinforcement for additional protection
- Internal fingertip reinforcement enhances abrasion resistance
- Synthetic leather palm is reinforced with touchscreen technology
- D3O™ palm padding absorbs impact and vibration
- Nylon carrier loops for convenient storage when not in use
- Ideal for military or law enforcement professionals
- EN 13594 rated impact protection
Specifications
Black | |
Specific | |
D3O™ Padding |
Hook and Loop | |
Synthetic Leather, Thermoplastic Rubber | |
MPT-55 |
WARNING: Cancer and Reproductive Harm -www.P65Warnings.ca.gov
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.