Learn More
Mechanix Wear™ M-Pact™ Fingerless Anti-Vibration Mechanics Gloves
Click to view available options
Medium, Large
Description
- Sonic welded thermoplastic rubber safegaurds knuckles
- Impact absorbing PORON™ XRD™ palm padding reduces shock and vibration
- Form-fitting TrekDry™ material keeps hands dry and comfortable
- Fingerless design allows for maximum control and dexterity
- Launderable
Specifications
Coating Material | Uncoated |
Color | Black |
Hand | Specific |
Material | Synthetic Leather |
Product Type | Mechanic's Gloves |
Size | Medium, Large |
Finger Style | Fingerless |
Glove Cuff Style | Elasticated |
Series | MFL-05 |