Mechanix Wear™ M-Pact Gloves

Full-fingered or fingerless—choose the style that best suits your needs.

Manufacturer: Mechanix Wear MPT02011

 View more versions of this product

Catalog No. 19-030-012D

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Finger Style Full
    Glove Cuff Style Shirred
    For Use With (Application) heavy equipment operation, towing and transportation, construction, maintenance and repair
    Material Synthetic Leather
    Size X-Large
    Color Red

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit