Mechanix Wear™ Original Gloves

Excellent all-purpose glove for mechanical and maintenance applications

Manufacturer: Mechanix Wear MG01011

 View more versions of this product

Catalog No. 19-813-589

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) military, law enforcement, firearm operation, maintenance repair operations
    Liner Material Foam
    Finger Style Full
    Exterior Finish Smooth
    Material Synthetic Leather, Clarino™, Thermal Plastic Rubber
    Size X-Large
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit