Learn More
MeCP2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259230
Catalog No. NBP259230
Add to cart
Description
Description
MeCP2 Polyclonal antibody specifically detects MeCP2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
MeCP2
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16
Rabbit
IgG
50 ug
Cancer, Chromatin Research, Neuroscience
Primary
4204
Western Blot
Western Blot 1:1000
Purified
MECP2
The exact immunogen is propietary information.
Protein G purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse