Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MECP2, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB31335

Catalog No. 89-998-384

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

MECP2
Unconjugated
In PBS (0.05% sodium azide, 0.05% proclin 300).
Liquid
MECP2
Recombinant protein corresponding to human MECP2.
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
methyl CpG binding protein 2 (Rett syndrome)
AUTSX3/DKFZp686A24160/MRX16/MRX79/MRXS13/MRXSL/PPMX/RTS/RTT
Rabbit
50 μg
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Primary
4204
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit