Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100608
Description
Antibody detects endogenous levels of total MED10.
MED10 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
Specifications
MED10 | |
Polyclonal | |
Unconjugated | |
MED10 | |
AA959813; AI385605; C78613; D13Wsu50e; L6; Med10; Mediator complex subunit 10; mediator of RNA polymerase II transcription subunit 10; mediator of RNA polymerase II transcription, subunit 10; mediator of RNA polymerase II transcription, subunit 10 homolog (NUT2, S. cerevisiae); NUT2; RGD1310128; Transformation-related gene 17 protein; transformation-related gene 20 protein; TRG17; TRG-17; TRG20; TRG-20 | |
Rabbit | |
Affinity chromatography | |
RUO | |
28077, 290939, 84246 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BTT4, Q9CXU0 | |
MED10 | |
A synthesized peptide derived from human MED10(Accession Q9BTT4), corresponding to amino acid residues E10-D60. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction