Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565847
Description
Immunogen sequence: MAEKFDHLEEH LEKFVENIRQ LGIIVSDFQP SSQAGLNQKL NFIVTGLQDI DKCRQQLHDI TVP Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
MED10 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
Specifications
MED10 | |
Polyclonal | |
Unconjugated | |
MED10 | |
AA959813; AI385605; C78613; D13Wsu50e; L6; Med10; Mediator complex subunit 10; mediator of RNA polymerase II transcription subunit 10; mediator of RNA polymerase II transcription, subunit 10; mediator of RNA polymerase II transcription, subunit 10 homolog (NUT2, S. cerevisiae); NUT2; RGD1310128; Transformation-related gene 17 protein; transformation-related gene 20 protein; TRG17; TRG-17; TRG20; TRG-20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84246 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTT4 | |
MED10 | |
Recombinant Human MED10. Recombinant protein control fragment (Product #RP-104990). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction