Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549894
Description
The antibody detects endogenous levels of total MED13 protein.
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. The product of this gene is proposed to form a sub-complex with MED12, cyclin C, and CDK8 that can negatively regulate transactivation by mediator. [provided by RefSeq].
Specifications
MED13 | |
Polyclonal | |
Unconjugated | |
MED13 | |
1110067M05Rik; activator-recruited cofactor 250 kDa component; ARC250; D030023K18; DRIP250; HSPC221; Kiaa0593; Med13; Mediator complex subunit 13; mediator of RNA polymerase II transcription subunit 13; mediator of RNA polymerase II transcription, subunit 13; mediator of RNA polymerase II transcription, subunit 13 homolog; Thrap1; thyroid hormone receptor associated protein 1; thyroid hormone receptor associated protein, 240 kDa subunit; thyroid hormone receptor-associated protein 1; Thyroid hormone receptor-associated protein complex 240 kDa component; thyroid hormone receptor-associated protein complex component TRAP240; thyroid hormone receptor-associated protein, 240 kDa subunit; Trap240; Vitamin D3 receptor-interacting protein complex component DRIP250; Zmediator complex subunit 13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9969 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UHV7 | |
MED13 | |
Synthesized peptide derived from internal of human MED13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction