Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED15 Mouse anti-Human, DyLight™ 550, Clone: OTI1C11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272618R
Description
MED15 Monoclonal specifically detects MED15 in Human samples. It is validated for Immunohistochemistry.Specifications
MED15 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MED15 | |
Recombinant protein fragment corresponding to amino acids 393-679 of human MED15 (NP_056973) produced in E.coli. | |
0.1 mL | |
51586.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1C11 | |
Immunohistochemistry | |
ARC105Arc105, CAG7A, CTG repeat protein 7a, CTG7A, FLJ42282, FLJ42935, mediator complex subunit 15PC2 glutamine/Q-rich-associated protein, mediator of RNA polymerase II transcription subunit 15, PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associatedprotein, PC2-glutamine-rich-associated protein, PCQAPDKFZp686A2214, Positive cofactor 2 glutamine/Q-rich-associated protein, positive cofactor 2, glutamine/Q-rich-associated protein, TIG1DKFZp762B1216, TIG-1TPA-inducible gene 1 protein, TNRC7, TPA inducible gene-1, TPA inducible protein, trinucleotide repeat containing 7,105-kD, Trinucleotide repeat-containing gene 7 protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title