Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCQAP Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549429

Catalog No. PA549429

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors. Required for cholesterol-dependent gene regulation. Positively regulates the Nodal signaling pathway.
Specifications

Specifications

PCQAP
Polyclonal
PBS with 0.09% sodium azide
Q924H2
Mediator of RNA polymerase II transcription subunit 15, Mediator complex subunit 15, Positive cofactor 2 glutamine/Q-rich-associated protein, PC2 glutamine/Q-rich-associated protein, mPcqap, Med15, Pcqap
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
360743, 94112
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Med15
Liquid
MED15
KLH conjugated synthetic peptide between 712-746 amino acids from the C-terminal region of mouse Med15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit