Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED19 Mouse anti-Human, Rat, Clone: OTI2E6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA810429

Description
MED19 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.Specifications
MED19 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
A0JLT2 | |
MED19 | |
Full length human recombit protein of human MED19 produced in E.coli. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
Western Blot | |
OTI2E6 | |
Unconjugated | |
MED19 | |
DT2P1G7, LCMR1, MED19AS | |
Mouse | |
Affinity Chromatography | |
RUO | |
219541, 311165 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title