Learn More
MED26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 210431AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3224
Add to Cart
Description
Description
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Specifications
Specifications
MED26
Polyclonal
Unconjugated
MED26
ARC70, CRSP complex subunit 7, CRSP7, CRSP70, MED26, mediator complex subunit 26
Rabbit
Antigen Affinity Chromatography
RUO
306328, 70625, 9441
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95402, Q7TN02
Med26
MED26 Fusion Protein Ag15381
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions