Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103919
Description
Antibody detects endogenous levels of total MED30.
Med30 is a subunit of the RNA polymerase II (Pol II) transcriptional mediator complex. The mediator complex is a coactivator involved in the regulated transcription of Pol II-dependent genes. The mediator complex functions as a bridge to convey information from gene-specific regulatory proteins to the basal Pol II transcription machinery, and is recruited to promoter regions by directly interacting with regulatory proteins. The mediator complex also serves as a scaffold for the assembly of a functional pre-initiation complex with Pol II and other general transcription factors. Found in the nucleus, Med30 is expressed in brain, heart, kidney, liver, lung, pancreas, placenta and skeletal muscle.
Specifications
MED30 | |
Polyclonal | |
Unconjugated | |
MED30 | |
1810038N03Rik; 2510044J04Rik; Med30; MED30S; Mediator complex subunit 30; mediator of RNA polymerase II transcription subunit 30; putative mediator of RNA polymerase II transcription subunit 30; THRAP6; thyroid hormone receptor associated protein 6; thyroid hormone receptor-associated protein 6; Thyroid hormone receptor-associated protein complex 25 kDa component; TRAP/Mediator complex component TRAP25; TRAP25 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
299905, 69790, 90390 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96HR3, Q9CQI9 | |
MED30 | |
A synthesized peptide derived from human MED30(Accession Q96HR3), corresponding to amino acid residues K98-K148. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction