Learn More
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159511AP
Catalog No. 50-173-0594
Add to cart
Description
Description
The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II (see MIM 180660)-transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (summary by Sato et al., 2004).
Specifications
Specifications
MED4
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q561Q8, Q9CQA5, Q9NPJ6
ARC36, DRIP36, HSPC126, MED4, mediator complex subunit 4, RP11 90M2.2, TRAP36, VDRIP
Rabbit
IgG
150 μL
-20°C
Primary
29079, 306030, 67381
Western Blot
Unconjugated
MED4
Liquid
MED4
MED4 Fusion Protein Ag8629
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat