Learn More
Aviva Systems Biology Corp MED7 Antibody - N-terminal region (ARP38445_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP38445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MED7. Alternate names: CRSP33, CRSP9, MGC12284, ARC34ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62991