Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen MED7
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125514
View Documents
Novus Biologicals
NBP31030625UL
25 μg
Each for $106.19
Add to cart
 
NB125513
View Documents
Novus Biologicals
NBP310306100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

MED7 Polyclonal antibody specifically detects MED7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

MED7
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS buffer, 2% sucrose
9443
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Human
Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33
The immunogen is a synthetic peptide directed towards the N terminal region of human MED7 (NP_004261). Peptide sequence MGEPQQVSALPPPPMQYIKEYTDENIQEGLAPKPPPPIKDSYMMFGNQFQ
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit