Medi-First™ Ibuprofen Tablets

Manufacturer: Moore Medical Corp 64096

 View more versions of this product

Catalog No. 19-898-323

  Check Availability
Add to Cart

    Specifications

    Active Ingredients Ibuprofen 200mg
    For Use With (Application) For headaches, muscular aches, pains and fever, minor arthritis and menstrual cramps
    Format Tablet
    Inactive Ingredients Carnauba Wax, Cellulose, Colloidal Silicon Dioxide, Corn Starch, Croscarmellose Sodium, Hypromellose, Iron Oxide red, Lactose, Magnesium Stearate, Microcrystalline Cellulose, Polydextrose, PEG, Polyvinyl Alcohol, Povidone, Pregelatinized Starch, Silica, Sodium Lauryl Sulfate, Sodium Starch Glycolate, Stearic Acid, Talc, Titanium Dioxide, Triacetate, Triacetin
    Quantity 250 Pack
    Storage Requirements 20 to 25°C
    Product Type Medi-First™ Ibuprofen Tablet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit