Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Capless

Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Quantity:
24 EA
48 EA

Clear Type I borosilicate glass bottle without graduations.

  • Manufactured from borosilicate glass which provides the best chemical resistance
  • Clear borosilicate glass bottle
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black phenolic screw cap can be purchased separately
Capacity (Metric) 125 mL
Color Clear
Material Borosilicate Glass (Type I)
Quantity 48 EA
Type Media Lab Bottle
Closure Size 33-430
Graduations NO
Height (Metric) 119 MM
Closure Type No cap