Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mediator of cell motility 1 Antibody (1E9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222593AF532
Description
mediator of cell motility 1 Monoclonal specifically detects mediator of cell motility 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
mediator of cell motility 1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MEMO1 | |
Recombinant human MEMO1 (1-297aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA | |
1000000000 | |
Western Blot, Flow Cytometry, ELISA | |
C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1 | |
Mouse | |
Protein G purified | |
Signal Transduction | |
51072.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction