Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mediator of cell motility 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15437320UL

 View more versions of this product

Catalog No. NBP15437320


Add to cart

Description

Description

Mediator of cell motility 1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

mediator of cell motility 1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
MEMO1
Synthetic peptides corresponding to MEMO1(mediator of cell motility 1) The peptide sequence was selected from the middle region of MEMO1. Peptide sequence AMESHKDEFTIIPVLVGALSESKEQEFGKLFSKYLADPSNLFVVSSDFCH.
34 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q9Y316
C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51072
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit