Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDICA 16, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
87272-20-6
C20H38O4
342.52
HYSMCRNFENOHJH-UHFFFAOYSA-N
medica 16,medic-16,unii-tc257flpck,tc257flpck,chembl63829,3,3,14,14-tetramethylhexanedecanedioic acid,3,3,14,14-tetramethyl-hexadecanedioic acid,hexadecandioic acid, 3,3,14,14-tetramethyl,medica16,d08lvh
121871
3,3,14,14-tetramethylhexadecanedioic acid
CC(C)(CCCCCCCCCCC(C)(C)CC(=O)O)CC(=O)O
This item is not returnable. View return policy
87272-20-6
C20H38O4
342.52
HYSMCRNFENOHJH-UHFFFAOYSA-N
medica 16,medic-16,unii-tc257flpck,tc257flpck,chembl63829,3,3,14,14-tetramethylhexanedecanedioic acid,3,3,14,14-tetramethyl-hexadecanedioic acid,hexadecandioic acid, 3,3,14,14-tetramethyl,medica16,d08lvh
121871
3,3,14,14-tetramethylhexadecanedioic acid
CC(C)(CCCCCCCCCCC(C)(C)CC(=O)O)CC(=O)O

FFA1 (GPR40) agonist; also inhibits ATP citrate lyase

Free fatty acid 1 (FFA1/GPR40) receptor agonist; exhibits selectivity for FFA1 (GPR40) over GPR120. Also inhibits ATP citrate lyase; reduces intracellular triacylglycerol levels in muscle, inhibits lipogenesis and increases insulin sensitivity in rodents in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 342.51
Percent Purity >98%
Chemical Name or Material MEDICA 16

Recommended Storage : Store at -20°C