Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medica Reference Electrodes for EasyBloodGas Analyzer

For use with EasyBloodGas Analyzer.

$356.50

Specifications

For Use With (Equipment) For easy blood gas analyzer
Quantity Each
Storage Requirements Room Temperature
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-515-405
View Documents
Medica
6204
Reference Electrode
Each for $356.50
Add to cart
 
Description

Description

For use with EasyBloodGas Analyzer.
Specifications

Specifications

For easy blood gas analyzer
Each
Room Temperature
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit