Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medica EasyLyte Analyzers

Completely automated, microprocessor-controlled electrolyte system uses ISE (Ion Selective Electrode) technology to make electrolyte measurements.

Manufacturer:  Medica 2005

 View more versions of this product

Catalog No. 22-515-352


Add to cart

Description

Description

 

  • CLIA Classification: Non-waived
  • Compact analyzer with small footprint.
  • Analysis time: blood, 60 seconds; urine†, 90 seconds
  • Completely automated, microprocessor-controlled with current ISE (Ion Selective Electrode) technology to make electrolyte measurements
  • Simple maintenance and replacement of electrodes, sample tube, and a simple pump tube
  • Calibrants packaged in a convenient solutions pack: 800mL solutions pack, for up to 1200 samples
  • Disposable, maintenance-free electrodes
  • Input normal ranges, quality control limits, and patient ID
  • All service calls can be performed by fax or telephone, eliminating the need for on-site service

  

  • Automatic or on-demand calibration
  • Measured and calculated results displayed and printed
  • Simple YES/NO numeric keypad menus, with alphanumeric display
  • Diagnostic software checks analyzer operation for quick and easy troubleshooting
  • Automatic probe-wiping ensures operator safety
  • RS-232 computer interface port
  • 40-column thermal printer

 

Specifications

Specifications

0.8A
8 in.
16.5 in.
50/60Hz
Whole Blood, Serum, Plasma or Urine
100 to 115VAC
9.5 in.
60 sec. (Blood) and 90 sec. (Urine)
Blood: 20 to 200mmol/L Na+, 0.2 to 40mmol/L K+, 0.2 to 5.0mmol/L Li+, Urine: 25 to 1000mmol/L (undiluted) Na+, 1 to 500mmol/L (undiluted) K+
0.10mmol/L Na+, 0.01mmol/L K+, 0.01mmol/L Li+
100 to 115VAC 50/60Hz
Yes
203mm
419mm
1:10
208 oz.
5.8kg
241mm
Measures various combinations of Sodium, Potassium, Lithium in whole Blood, serum, plasma or urine
≤85%
100mL Whole Blood, serum or plasma, 60mL capillary sample, 400mL diluted (1:10) urine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit