Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medica EasyLyte Analyzers
Completely automated, microprocessor-controlled electrolyte system uses ISE (Ion Selective Electrode) technology to make electrolyte measurements.
$9210.00 - $9690.00
Specifications
Amperage | 0.8A |
---|---|
Analysis Time | 60 sec. (Blood) and 90 sec. (Urine) |
Calibration | Yes |
Depth (English) | 8 in. |
Depth (Metric) | 203mm |
Catalog Number | Mfr. No. | Sample Type | Applications | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Type | Applications | Price | Quantity & Availability | ||||
22-515-352
|
Medica
2005 |
Whole Blood, Serum, Plasma or Urine | Measures various combinations of Sodium, Potassium, Lithium in whole Blood, serum, plasma or urine |
Each for $9,690.00
|
|
22-515-351
|
Medica
2012 |
1:10 (urine) | Measures various combinations of Sodium, Potassium, Chloride in whole Blood, serum, plasma or urine |
Each for $9,210.00
|
|
Description
- CLIA Classification: Non-waived
- Compact analyzer with small footprint.
- Analysis time: blood, 60 seconds; urine, 90 seconds
- Completely automated, microprocessor-controlled with current ISE (Ion Selective Electrode) technology to make electrolyte measurements
- Simple maintenance and replacement of electrodes, sample tube, and a simple pump tube
- Calibrants packaged in a convenient solutions pack: 800mL solutions pack, for up to 1200 samples
- Disposable, maintenance-free electrodes
- Input normal ranges, quality control limits, and patient ID
- All service calls can be performed by fax or telephone, eliminating the need for on-site service
- Automatic or on-demand calibration
- Measured and calculated results displayed and printed
- Simple YES/NO numeric keypad menus, with alphanumeric display
- Diagnostic software checks analyzer operation for quick and easy troubleshooting
- Automatic probe-wiping ensures operator safety
- RS-232 computer interface port
- 40-column thermal printer
Specifications
0.8A | |
Yes | |
203mm | |
419mm | |
100mL Whole Blood, serum or plasma, 60mL capillary sample, 400mL diluted (1:10) urine | |
208 oz. | |
9.5 in. |
60 sec. (Blood) and 90 sec. (Urine) | |
8 in. | |
16.5 in. | |
≤85% | |
100 to 115VAC | |
5.8kg | |
241mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title