Medica EasyLyte Analyzers

Compact analyzer with small footprint measures various combinations of sodium, potassium, chloride, lithium, calcium, and pH in whole blood, serum, plasma, or urine†

Manufacturer: Medica CorporationSupplier Diversity Partner 2012

 View more versions of this product

Catalog No. 22-515-351

  Check Availability
Add to Cart

    Specifications

    Height (Metric) 419mm
    Measurement Method Direct measurement by ISE
    Sample Dilution 1:10
    Amperage 0.8A
    Sample Type 1:10 (urine)
    Weight (English) 208 oz.
    Voltage 100 to 115VAC
    Weight (Metric) 5.8kg
    Width (English) 9.5 in.
    Width (Metric) 241mm
    Analysis Time 60 sec. (Blood) and 90 sec. (Urine)
    Applications Measures various combinations of Sodium, Potassium, Chloride in whole Blood, serum, plasma or urine
    Resolution 0.10mmol/L Na+, 0.01mmol/L K+, 0.10mmol/L Cl-
    Sample Size 100mL Whole Blood, serum or plasma, 60mL capillary sample, 400mL diluted (1:10) urine
    Calibration Yes
    Depth (English) 8 in.
    Depth (Metric) 203mm
    Height (English) 16.5 in.
    Detection Range Blood: 20 to 200mmol/L Na+, 0.2 to 40mmol/L K+, 25 to 200 mmol/L Cl-, Urine: 25 to 1000mmol/L (undiluted) Na+, 1 to 500mmol/L (undiluted) K+, 25 to 500mmol/L (undiluted) Cl-
    Humidity ≤85%

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit