Medical Action™ Autoclave Bags

Manufacturer: American Precision Plastics 848

 View more versions of this product

Catalog No. 14-828-144

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Includes Indicator, flat pack, twist ties, biohazard labeling, biohazard symbol
    Thickness 1.8mil
    Autoclavable Yes
    Disposable Yes
    Max. Temperature (English) 285°F
    Max. Temperature (Metric) 140.5°C
    Capacity (English) 44 gal.
    Capacity (Metric) 166.55L
    Material Polypropylene
    Color Red, Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit