Medical Action Biohazardous Waste Bags, LLDPE Film, Flat Pack Available on GSA/VA Contract for Federal Government customers only.

Passes ASTMD1709-85, 165 Gram Dart Test

Manufacturer: American Precision Plastics HRD242411

 View more versions of this product

Catalog No. 14-828-186

  Check Availability
Add to Cart

    Specifications

    Product Type Biohazardous Waste Bag
    Includes Biohazardous or infectious waste with biohazard symbol
    Material LLDPE
    Thickness (English) 0.433 in.
    Thickness 11 mic
    Length (English) 24 in.
    Width (Metric) 60.96cm
    Capacity 8 to 10 gal., 30.28 to 37.87L
    Dimensions (L x W) 24 x 24 in. (60.96 x 60.96cm)
    Capacity (English) 8 to 10 gal.
    Capacity (Metric) 30.28 to 37.87L
    Certifications/Compliance ASTM Dart
    For Use With (Application) Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab
    Thickness (Metric) 11mm
    Color Red, Black
    Length (Metric) 60.96cm
    Width (English) 24 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit