Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
These Biohazardous Waste Bags pass ASTMD1709-85, 165 Gram Dart Test.
Supplier: Medegen 119
Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Biohazardous Waste Bag | |
113.6 L | |
Red, Black | |
Biohazardous or infectious waste with biohazard symbol | |
0.076 mm | |
30.5 x 41 in. (77.47 x 104.14cm) | |
77.47 cm | |
104.14 cm |
30 gal. | |
ASTM Dart | |
Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab | |
LLDPE | |
3 mil | |
30.5 in. | |
41 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
May NOT be used in CA, FL, PA or for interstate (between state) transport.
Spot an opportunity for improvement?
Provide Content Correction