Learn More
Medical Action™ High-Density Institutional Trash Can Liners Available on GSA/VA Contract for Federal Government customers only.
High density trash can liners for use in hospitals, elder care centers, and other institutional settings
Manufacturer:  American Precision Plastics HCL386016
 View more versions of this product
Catalog No. 14-828-181
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
38 x 60 in. (96.5 x 152.4cm)
38 in.
16mic
60 gal.
Clear
HDPE (High-density Polyethylene)
Hospitals, elder care centers and other institutional settings
Waste Disposal Bag
60 in.
227L
96.5cm
152.4cm